Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122667.2 | NP_001116139.1 | 4 - 5 | 386 | 101 | |
NM_153588.3 | NP_705816.2 | 2 - 3 | 400 | 101 | ||
GenBank mRNA | AF532598.1 | - | 2 - 3 | 182 | 101 | |
AK220377.1 | - | 2 - 3 | 182 | 101 | ||
BC131644.1 | - | 2 - 3 | 182 | 40110019 | 101 | |
BC131645.1 | - | 2 - 3 | 180 | 101 | ||
CJ126058.1 | - | 2 - 3 | 402 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |