Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122667.2 | NP_001116139.1 | 13 - 14 | 2550 | 69 | |
NM_153588.3 | NP_705816.2 | 11 - 12 | 2564 | 69 | ||
GenBank mRNA | AF532598.1 | - | 11 - 12 | 2346 | 69 | |
AK220377.1 | - | 11 - 12 | 2346 | 69 | ||
BC131644.1 | - | 11 - 12 | 2346 | 40110019 | 69 | |
BC131645.1 | - | 11 - 12 | 2344 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |