Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170591.1 | NP_733479.1 | 12 - 13 | 1509 | 75 | |
GenBank mRNA | AK006062.1 | - | 12 - 13 | 1612 | 75 | |
AK029688.1 | - | 1563 | 75 | |||
AK078279.1 | - | 12 - 13 | 1557 | 75 | ||
AK086412.1 | - | 12 - 13 | 1577 | 75 | ||
AK129134.1 | - | 12 - 13 | 1507 | 75 | ||
AK133721.1 | - | 12 - 13 | 1520 | 75 | ||
AK139895.1 | - | 12 - 13 | 1640 | 75 | ||
AK146477.1 | - | 12 - 13 | 1509 | 75 | ||
AK200201.1 | - | 2 - 3 | 237 | 75 | ||
BC026743.1 | - | 12 - 13 | 1540 | 3964978 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |