Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 3 - 4 | 800 | 91 | |
NM_009765.3 | NP_033895.2 | 3 - 4 | 606 | 91 | ||
GenBank mRNA | AK145017.1 | - | 3 - 4 | 767 | 91 | |
AK148272.1 | - | 3 - 4 | 604 | 91 | ||
AK164598.1 | - | 3 - 4 | 596 | 91 | ||
U65594.1 | - | 3 - 4 | 583 | 91 | ||
U72947.1 | - | 2 - 3 | 242 | 91 | ||
U82270.1 | - | 2 - 3 | 292 | 91 | ||
U89652.1 | - | 2 - 3 | 330 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons |