Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205343.1 | NP_001192272.1 | 2 - 3 | 143 | 77 | |
NM_130887.3 | NP_570957.2 | 2 - 3 | 143 | 77 | ||
GenBank mRNA | AF314171.1 | - | 2 - 3 | 116 | 77 | |
AK053198.1 | - | 2 - 3 | 144 | 77 | ||
AK086767.1 | - | 2 - 3 | 124 | 77 | ||
AK138980.1 | - | 2 - 3 | 116 | 77 | ||
BB664306.1 | - | 2 - 3 | 144 | 77 | ||
BC132475.1 | - | 2 - 3 | 83 | 40130752 | 77 | |
BC137854.1 | - | 2 - 3 | 83 | 8860873 | 77 | |
BC144975.1 | - | 2 - 3 | 83 | 9053523 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |