Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133768.4 | NP_598529.1 | 10 - 11 | 796 | 63 | |
GenBank mRNA | AK076453.1 | - | 10 - 11 | 811 | 63 | |
AK138423.1 | - | 10 - 11 | 793 | 63 | ||
AK140160.1 | - | 10 - 11 | 811 | 63 | ||
AK148764.1 | - | 10 - 11 | 780 | 63 | ||
AK168739.1 | - | 10 - 11 | 791 | 63 | ||
AK187224.1 | - | 1 - 2 | 79 | 63 | ||
BC016670.1 | - | 10 - 11 | 796 | 2648753 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |