Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133768.4 | NP_598529.1 | 12 - 13 | 998 | 58 | |
GenBank mRNA | AK010922.1 | - | 12 - 13 | 1010 | 58 | |
AK076453.1 | - | 12 - 13 | 1013 | 58 | ||
AK138423.1 | - | 12 - 13 | 995 | 58 | ||
AK140160.1 | - | 12 - 13 | 1013 | 58 | ||
AK148764.1 | - | 12 - 13 | 982 | 58 | ||
AK168739.1 | - | 12 - 13 | 993 | 58 | ||
BC016670.1 | - | 12 - 13 | 998 | 2648753 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |