Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290747.1 | NP_001277676.1 | 3 - 4 | 574 | 142 | |
NM_145224.3 | NP_660259.1 | 6 - 7 | 831 | 142 | ||
NM_181319.5 | NP_851836.2 | 6 - 7 | 935 | 142 | ||
GenBank mRNA | AF516208.1 | - | 6 - 7 | 935 | 142 | |
AK053430.1 | - | 3 - 4 | 451 | 142 | ||
AK084312.1 | - | 6 - 7 | 832 | 142 | ||
AY125891.1 | - | 5 - 6 | 787 | 142 | ||
BC152402.1 | - | 3 - 4 | 574 | 40140811 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |