Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172053.2 | NP_742050.2 | 5 - 6 | 1220 | 90 | |
GenBank mRNA | AK031314.1 | - | 5 - 6 | 1186 | 90 | |
AK173338.1 | - | 5 - 6 | 1156 | 90 | ||
BC034739.1 | - | 5 - 6 | 1233 | 4951547 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |