Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 29 - 30 | 5098 | 88 | |
GenBank mRNA | AF300701.1 | - | 28 - 29 | 4575 | 88 | |
AK156112.1 | - | 29 - 30 | 5101 | 88 | ||
U36488.1 | - | 25 - 26 | 4260 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons |