Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpzl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007962.4 | NP_031988.2 | 1 - 2 | 455 | 69 | |
GenBank mRNA | AF030454.1 | - | 1 - 2 | 236 | 69 | |
AK032788.1 | - | 1 - 2 | 236 | 69 | ||
AK166341.1 | - | 1 - 2 | 186 | 69 | ||
AK167263.1 | - | 1 - 2 | 219 | 69 | ||
AK170816.1 | - | 1 - 2 | 221 | 69 | ||
BC015076.1 | - | 1 - 2 | 281 | 4188907 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |