Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143616.1 | - | 11 - 12 | 1509 | 79 | |
AK144669.1 | - | 11 - 12 | 1516 | 79 | ||
AK146813.1 | - | 11 - 12 | 1478 | 79 | ||
AK146918.1 | - | 11 - 12 | 1486 | 79 | ||
BC003741.1 | - | 11 - 12 | 1475 | 3488803 | 79 | |
D63850.1 | - | 11 - 12 | 1492 | 79 | ||
FN687734.2 | - | 11 - 12 | 1235 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |