Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019986.3 | NP_064370.2 | 2 - 3 | 638 | 70 | |
GenBank mRNA | AF227684.1 | - | 2 - 3 | 581 | 70 | |
AK016800.1 | - | 2 - 3 | 681 | 70 | ||
AK144206.1 | - | 2 - 3 | 622 | 70 | ||
AK146525.1 | - | 2 - 3 | 611 | 70 | ||
BC082806.1 | - | 2 - 3 | 640 | 70 | ||
BC119460.2 | - | 1 - 2 | 135 | 70 | ||
BC148304.2 | - | 1 - 2 | 142 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |