Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021292.2 | NP_067267.2 | 18 - 19 | 2772 | 100 | |
GenBank mRNA | AJ250841.1 | - | 18 - 19 | 2752 | 100 | |
AK039759.1 | - | 18 - 19 | 2743 | 100 | ||
AK148262.1 | - | 18 - 19 | 2745 | 100 | ||
AK161910.1 | - | 5 - 6 | 1422 | 100 | ||
JQ599253.1 | - | 2 - 3 | 208 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |