Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023508.6 | NP_075997.1 | 3 - 4 | 309 | 107 | |
GenBank mRNA | AK005602.1 | - | 3 - 4 | 332 | 107 | |
AK005814.1 | - | 3 - 4 | 309 | 107 | ||
AK006040.1 | - | 2 - 3 | 211 | 107 | ||
AK006773.1 | - | 3 - 4 | 326 | 107 | ||
AK132738.1 | - | 3 - 4 | 356 | 107 | ||
AK133140.1 | - | 2 - 3 | 172 | 107 | ||
BC048432.1 | - | 3 - 4 | 311 | 6773342 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |