Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023622.4 | NP_076111.2 | 13 - 14 | 1536 | 83 | |
GenBank mRNA | AF237669.1 | - | 13 - 14 | 1495 | 83 | |
AF239661.1 | - | 13 - 14 | 1537 | 83 | ||
AK004876.1 | - | 10 - 11 | 1565 | 83 | ||
AK053550.1 | - | 13 - 14 | 1523 | 83 | ||
AK131140.1 | - | 1042 | 83 | |||
AK165023.1 | - | 10 - 11 | 1925 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |