Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023712.3 | NP_076201.2 | 9 - 10 | 2083 | 66 | |
NR_045537.1 | 10 - 11 | 2062 | 66 | |||
GenBank mRNA | AF212372.1 | - | 9 - 10 | 1574 | 66 | |
AK008721.1 | - | 10 - 11 | 1559 | 66 | ||
AK033907.1 | - | 7 - 8 | 2638 | 66 | ||
AK154353.1 | - | 9 - 10 | 1573 | 66 | ||
AK166931.1 | - | 9 - 10 | 1575 | 66 | ||
BC002297.1 | - | 9 - 10 | 1102 | 66 | ||
BC026854.1 | - | 9 - 10 | 1544 | 4242049 | 66 | |
BC085491.1 | - | 9 - 10 | 1527 | 6829415 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |