Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023712.3 | NP_076201.2 | 10 - 11 | 2171 | 107 | |
NR_045537.1 | 11 - 12 | 2150 | 107 | |||
GenBank mRNA | AF212372.1 | - | 10 - 11 | 1662 | 107 | |
AK008721.1 | - | 11 - 12 | 1647 | 107 | ||
AK033907.1 | - | 8 - 9 | 2726 | 107 | ||
AK154353.1 | - | 10 - 11 | 1661 | 106 | ||
AK166931.1 | - | 10 - 11 | 1663 | 107 | ||
BC002297.1 | - | 10 - 11 | 1190 | 107 | ||
BC026854.1 | - | 10 - 11 | 1632 | 4242049 | 107 | |
BC085491.1 | - | 10 - 11 | 1615 | 6829415 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |