Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026302.3 | NP_080578.1 | 10 - 11 | 976 | 80 | |
GenBank mRNA | AK016059.1 | - | 10 - 11 | 1030 | 80 | |
AK034292.1 | - | 11 - 12 | 1034 | 80 | ||
AK141855.1 | - | 11 - 12 | 999 | 80 | ||
AK160813.1 | - | 10 - 11 | 982 | 80 | ||
AK163240.1 | - | 10 - 11 | 998 | 80 | ||
AK168484.1 | - | 10 - 11 | 982 | 80 | ||
BC006677.1 | - | 10 - 11 | 939 | 80 | ||
BC034725.1 | - | 10 - 11 | 976 | 3667031 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |