Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026539.2 | NP_080815.1 | 17 - 18 | 2053 | 100 | |
GenBank mRNA | AK014473.1 | - | 17 - 18 | 2055 | 100 | |
AK165656.1 | - | 17 - 18 | 2080 | 100 | ||
BC020486.1 | - | 2 - 3 | 225 | 100 | ||
BC052385.1 | - | 17 - 18 | 2037 | 5703859 | 100 | |
BC057567.1 | - | 17 - 18 | 2066 | 6415898 | 100 | |
BC062966.1 | - | 17 - 18 | 2066 | 6415898 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |