Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029814.1 | NP_084090.1 | 6 - 7 | 546 | 85 | |
GenBank mRNA | AK003313.1 | - | 1 - 2 | 295 | 85 | |
AK008911.1 | - | 6 - 7 | 546 | 85 | ||
AK147058.1 | - | 6 - 7 | 568 | 85 | ||
BC006947.1 | - | 6 - 7 | 507 | 2811290 | 85 | |
BC080673.1 | - | 5 - 6 | 1341 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |