Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080819.1 | NP_001074288.1 | 7 - 8 | 2424 | 69 | |
GenBank mRNA | AF268912.1 | - | 6 - 7 | 1275 | 69 | |
AK142143.1 | - | 6 - 7 | 1606 | 69 | ||
BC082554.1 | - | 1656 | 30622048 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |