Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053250.2 | NP_444480.1 | 2 - 3 | 152 | 83 | |
NM_181664.2 | NP_858050.1 | 2 - 3 | 152 | 83 | ||
GenBank mRNA | AF367970.1 | - | 2 - 3 | 152 | 83 | |
AF367971.1 | - | 2 - 3 | 152 | 83 | ||
AK080272.1 | - | 2 - 3 | 152 | 83 | ||
AY555745.1 | - | 2 - 3 | 143 | 83 | ||
BC116360.1 | - | 2 - 3 | 152 | 40099973 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |