Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB036742.1 | - | 1 | 552 | 65 | |
BC125442.1 | - | 1 | 320 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |