Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199347.1 | NP_001186276.1 | 4 - 5 | 554 | 81 | |
NM_001199348.1 | NP_001186277.1 | 3 - 4 | 412 | 81 | ||
NM_054039.2 | NP_473380.1 | 3 - 4 | 487 | 81 | ||
GenBank mRNA | AF277991.1 | - | 3 - 4 | 412 | 81 | |
AF277992.1 | - | 3 - 4 | 470 | 81 | ||
AK157447.1 | - | 4 - 5 | 555 | 81 | ||
AY357712.1 | - | 2 - 3 | 270 | 81 | ||
AY357713.1 | - | 2 - 3 | 270 | 81 | ||
BC132333.1 | - | 2 - 3 | 258 | 40130610 | 81 | |
BC132335.1 | - | 2 - 3 | 258 | 40130612 | 81 | |
DQ387959.1 | - | 4 - 5 | 554 | 81 | ||
KF471319.1 | - | 2 - 3 | 254 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |