Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267625.1 | NP_001254554.1 | 6 - 7 | 638 | 61 | |
NM_001267626.1 | NP_001254555.1 | 6 - 7 | 638 | 61 | ||
NM_058212.2 | NP_478119.1 | 6 - 7 | 635 | 61 | ||
GenBank mRNA | AF362750.1 | - | 6 - 7 | 607 | 61 | |
AK012970.1 | - | 6 - 7 | 643 | 61 | ||
AK039011.1 | - | 6 - 7 | 619 | 61 | ||
BC048572.1 | - | 6 - 7 | 518 | 6705809 | 61 | |
FJ151218.1 | - | 6 - 7 | 610 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |