Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270996.1 | NP_001257925.1 | 10 - 11 | 1132 | 87 | |
NM_007404.2 | NP_031430.2 | 10 - 11 | 1132 | 87 | ||
GenBank mRNA | AK122188.1 | - | 10 - 11 | 1115 | 87 | |
AK148174.1 | - | 10 - 11 | 1018 | 87 | ||
AK155672.1 | - | 10 - 11 | 1025 | 87 | ||
AK161778.1 | - | 10 - 11 | 1056 | 87 | ||
BC047156.1 | - | 10 - 11 | 1010 | 6488707 | 87 | |
BC052710.1 | - | 10 - 11 | 1004 | 87 | ||
U41765.1 | - | 10 - 11 | 1004 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |