Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007422.3 | NP_031448.2 | 3 - 4 | 473 | 86 | |
GenBank mRNA | AK004877.1 | - | 3 - 4 | 426 | 86 | |
AK010263.1 | - | 3 - 4 | 448 | 86 | ||
AK028060.1 | - | 3 - 4 | 473 | 86 | ||
AK043273.1 | - | 3 - 4 | 466 | 86 | ||
AK148420.1 | - | 3 - 4 | 455 | 86 | ||
AK170279.1 | - | 3 - 4 | 459 | 86 | ||
BC139404.1 | - | 3 - 4 | 385 | 8862426 | 86 | |
BC139417.1 | - | 3 - 4 | 385 | 8862439 | 86 | |
L24554.1 | - | 3 - 4 | 440 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |