Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200011094.mm.1 |
NM_001164090.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011094.mm.1 |
NM_001164090.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011094.mm.1 |
TC1200001597.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011090.mm.1 |
NM_001164090.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011090.mm.1 |
NM_001164090.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011090.mm.1 |
NM_007530.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011090.mm.1 |
NM_007530.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011090.mm.1 |
TC1200001597.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011093.mm.1 |
NM_007530.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011093.mm.1 |
NM_007530.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011093.mm.1 |
TC1200001597.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011091.mm.1 |
NM_001164090.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011091.mm.1 |
NM_001164090.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011091.mm.1 |
NM_007530.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011091.mm.1 |
NM_007530.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011091.mm.1 |
TC1200001597.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011092.mm.1 |
NM_007530.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011092.mm.1 |
NM_007530.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011092.mm.1 |
TC1200001597.mm.1 |
MTA TC |