Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008486.2 | NP_032512.2 | 14 - 15 | 2254 | 121 | |
GenBank mRNA | AK143746.1 | - | 14 - 15 | 2197 | 121 | |
AK171427.1 | - | 15 - 16 | 2616 | 121 | ||
BC005431.1 | - | 14 - 15 | 2220 | 3593307 | 121 | |
BC017011.1 | - | 14 - 15 | 2278 | 3590117 | 121 | |
BC040792.1 | - | 14 - 15 | 2278 | 3590117 | 121 | |
U77083.1 | - | 14 - 15 | 2186 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |