Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476300_m1
          See other NCF4 GE Assays ›
          Gene Symbol:
          Ncf4
          Gene Name:
          neutrophil cytosolic factor 4
          Gene Aliases:
          AI451400, p40phox
          Chromosome Location:
          Chr.15: 78244164 - 78262582 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476300_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ncf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008677.2 NP_032703.2 3 - 4 512 71
          GenBank mRNA AB002665.1 - 3 - 4 489 71
          AK150510.1 - 3 - 4 494 71
          AK171315.1 - 3 - 4 290 71
          BC025517.1 - 3 - 4 341 5321182 71
          FJ168470.1 - 3 - 4 303 71
          U59488.1 - 3 - 4 459 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000640.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ncf4

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of catalytic activity
          protein binding
          lipid binding
          superoxide-generating NADPH oxidase activator activity
          phosphatidylinositol-3-phosphate binding
          phosphatidylinositol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 AK171315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 BC025517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 U59488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 NM_008677.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 AA016555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 AK171315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 BC025517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 U59488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 NM_008677.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377452 AA016555.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37839 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170385 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106935 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 AK171315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 BC025517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 U59488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 NM_008677.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 AA016555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 AK171315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 BC025517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 U59488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 NM_008677.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377452 AA016555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 AK171315.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 BC025517.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 U59488.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 NM_008677.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 AA016555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 AK171315.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 BC025517.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 U59488.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 NM_008677.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377452 AA016555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 AK171315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 BC025517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 U59488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 NM_008677.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 AA016555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 AK171315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 BC025517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 U59488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 NM_008677.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377452 AA016555.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107398 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11914382 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11914384 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11914370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11914390 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11914378 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11914376 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425061 - -
          Affymetrix HT MG-430 PM Array Plate 1418465_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425053 - -
          Mouse Genome 430 2.0 Array 1418465_at - -
          Mouse Genome 430 2.0 Array 1418465_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17972_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17972_at - -
          Affymetrix Mouse Clariom S Array PSR1500004638.mm.1 NM_008677.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004638.mm.1 NM_008677.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004638.mm.1 TC1500000640.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004639.mm.1 NM_008677.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004639.mm.1 NM_008677.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004639.mm.1 TC1500000640.mm.1 MTA TC
          Mouse Expression Set 430 1418465_PM_at - -
          Mouse Expression Set 430 1418465_at - -
          Murine Genome U74v2 Set 103662_PM_at - -
          Murine Genome U74v2 Set 103662_at - -
          Mouse Genome 430A 2.0 Array 1418465_PM_at - -
          Mouse Genome 430A 2.0 Array 1418465_at - -
          Affymetrix HT Mouse Genome 430A Array 1418465_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418465_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 919748 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 610164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2743013 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2743013 - -
          mouse-6 v1.1 (Illumina) scl47772.10.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 610164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47772.10.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610164 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2743013 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2743013 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl47772.10.1_9-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0