Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000019108.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019108.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019108.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019108.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019108.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019109.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019109.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019109.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019109.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019109.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019110.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019110.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019110.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019110.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019110.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019106.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019106.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019106.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019106.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019106.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019107.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019107.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019107.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019107.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019107.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019112.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019112.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019112.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019112.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019112.mm.1 |
TC1000002541.mm.1 |
MTA TC |