Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163662.1 | NP_001157134.1 | 9 - 10 | 1007 | 65 | |
NM_008749.2 | NP_032775.1 | 9 - 10 | 1007 | 65 | ||
GenBank mRNA | AK002630.1 | - | 9 - 10 | 988 | 65 | |
AK004886.1 | - | 9 - 10 | 978 | 65 | ||
AK007976.1 | - | 9 - 10 | 981 | 65 | ||
AK045087.1 | - | 9 - 10 | 1007 | 65 | ||
AK140710.1 | - | 9 - 10 | 985 | 65 | ||
AK160981.1 | - | 9 - 10 | 994 | 65 | ||
AK164088.1 | - | 9 - 10 | 989 | 65 | ||
AK170717.1 | - | 9 - 10 | 1002 | 65 | ||
BC072554.1 | - | 9 - 10 | 985 | 5684979 | 65 | |
M96823.1 | - | 8 - 9 | 929 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |