Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008851.4 | NP_032877.1 | 20 - 21 | 3721 | 88 | |
NR_075078.1 | 21 - 22 | 3415 | 88 | |||
GenBank mRNA | AF006467.1 | - | 21 - 22 | 3296 | 88 | |
AK155377.1 | - | 20 - 21 | 4194 | 88 | ||
BC044893.1 | - | 20 - 21 | 3226 | 4481550 | 88 | |
BC048150.1 | - | 20 - 21 | 3375 | 5706382 | 88 | |
Y08922.1 | - | 20 - 21 | 3722 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |