Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008921.2 | NP_032947.1 | 11 - 12 | 1164 | 64 | |
GenBank mRNA | AK011456.1 | - | 11 - 12 | 1146 | 64 | |
AK088557.1 | - | 11 - 12 | 1184 | 64 | ||
AK089063.1 | - | 11 - 12 | 1189 | 64 | ||
AK133878.1 | - | 11 - 12 | 1161 | 64 | ||
AK160831.1 | - | 9 - 10 | 2926 | 64 | ||
BC089544.1 | - | 11 - 12 | 1203 | 30504228 | 64 | |
D13544.1 | - | 11 - 12 | 1164 | 64 | ||
J04620.1 | - | 11 - 12 | 1223 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |