Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164677.1 | NP_001158149.1 | 4 - 5 | 607 | 108 | |
NM_001164678.1 | NP_001158150.1 | 4 - 5 | 607 | 108 | ||
NM_011052.2 | NP_035182.2 | 4 - 5 | 607 | 108 | ||
GenBank mRNA | AF119955.1 | - | 4 - 5 | 568 | 108 | |
AJ005073.1 | - | 4 - 5 | 455 | 108 | ||
AK029827.1 | - | 4 - 5 | 574 | 108 | ||
AK031256.1 | - | 4 - 5 | 593 | 108 | ||
AK129340.1 | - | 4 - 5 | 577 | 108 | ||
AK145893.1 | - | 4 - 5 | 595 | 108 | ||
AK147736.1 | - | 4 - 5 | 580 | 108 | ||
AK167574.1 | - | 4 - 5 | 565 | 108 | ||
AK169704.1 | - | 4 - 5 | 608 | 108 | ||
BC026823.1 | - | 4 - 5 | 549 | 4038684 | 108 | |
BC051123.1 | - | 4 - 5 | 458 | 6529850 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |