Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600018210.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018210.mm.1 |
NM_001159953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018210.mm.1 |
NM_001159953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_001159953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_001159953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_001159955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_001159955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_001159956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_001159956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_011054.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018213.mm.1 |
NM_011054.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001025568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001025568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_001159960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_011054.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018219.mm.1 |
NM_011054.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018211.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018211.mm.1 |
NM_001159955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018211.mm.1 |
NM_001159955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018211.mm.1 |
NM_001159956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018211.mm.1 |
NM_001159956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018211.mm.1 |
NM_011054.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018211.mm.1 |
NM_011054.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018212.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018212.mm.1 |
NM_001159955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018212.mm.1 |
NM_001159955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018212.mm.1 |
NM_001159956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018212.mm.1 |
NM_001159956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018212.mm.1 |
NM_011054.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018212.mm.1 |
NM_011054.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001025568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001025568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_001159960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_011054.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018220.mm.1 |
NM_011054.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001025568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001025568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001159952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001159952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001159957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001159957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001159960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018217.mm.1 |
NM_001159960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001025568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001025568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001159952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001159952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001159957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001159957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001159960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018218.mm.1 |
NM_001159960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018209.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018209.mm.1 |
NM_001159953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018209.mm.1 |
NM_001159953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
TC0600002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001025568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001025568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001159952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001159952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001159957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001159957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001159960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018216.mm.1 |
NM_001159960.1 |
Refseq |