Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142916.1 | NP_001136388.1 | 17 - 18 | 2084 | 98 | |
NM_011961.3 | NP_036091.2 | 16 - 17 | 2021 | 98 | ||
GenBank mRNA | AF080572.1 | - | 16 - 17 | 1792 | 98 | |
AK033363.1 | - | 17 - 18 | 2043 | 98 | ||
AK046364.1 | - | 16 - 17 | 2025 | 98 | ||
AK157207.1 | - | 17 - 18 | 2044 | 98 | ||
BC021352.1 | - | 16 - 17 | 1853 | 5007509 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |