Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013464.4 | NP_038492.1 | 1 - 2 | 427 | 105 | |
GenBank mRNA | AF405560.1 | - | 1 - 2 | 61 | 105 | |
AF405561.1 | - | 1 - 2 | 61 | 105 | ||
AF405562.1 | - | 1 - 2 | 61 | 105 | ||
AF405563.1 | - | 1 - 2 | 61 | 105 | ||
AF405565.1 | - | 1 - 2 | 61 | 105 | ||
AF405566.1 | - | 1 - 2 | 61 | 105 | ||
AF405567.1 | - | 1 - 2 | 61 | 105 | ||
AF405568.1 | - | 1 - 2 | 61 | 105 | ||
AF405570.1 | - | 1 - 2 | 61 | 105 | ||
AK153678.1 | - | 1 - 2 | 428 | 105 | ||
AK155380.1 | - | 1 - 2 | 479 | 105 | ||
AY662080.1 | - | 1 - 2 | 61 | 105 | ||
AY662081.1 | - | 1 - 2 | 61 | 105 | ||
AY662082.1 | - | 1 - 2 | 61 | 105 | ||
AY662083.1 | - | 1 - 2 | 61 | 105 | ||
AY662084.1 | - | 1 - 2 | 61 | 105 | ||
D38416.1 | - | 1 - 2 | 76 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |