Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285833.1 | NP_001272762.1 | 7 - 8 | 737 | 108 | |
NM_001285835.1 | NP_001272764.1 | 7 - 8 | 737 | 108 | ||
NM_015760.5 | NP_056575.1 | 7 - 8 | 741 | 108 | ||
GenBank mRNA | AB041034.1 | - | 7 - 8 | 592 | 108 | |
AB042745.1 | - | 7 - 8 | 592 | 108 | ||
AF218723.1 | - | 7 - 8 | 592 | 108 | ||
AF261944.1 | - | 7 - 8 | 596 | 108 | ||
AF276957.1 | - | 7 - 8 | 603 | 108 | ||
AK050371.1 | - | 7 - 8 | 596 | 108 | ||
AK052905.1 | - | 7 - 8 | 592 | 108 | ||
AK085509.1 | - | 7 - 8 | 597 | 108 | ||
AK149398.1 | - | 7 - 8 | 592 | 108 | ||
AK149455.1 | - | 7 - 8 | 592 | 108 | ||
AK149492.1 | - | 7 - 8 | 592 | 108 | ||
BC021378.1 | - | 7 - 8 | 585 | 108 | ||
BC138918.1 | - | 7 - 8 | 588 | 8861940 | 108 | |
BC145462.1 | - | 7 - 8 | 584 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |