Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015765.2 | NP_056580.2 | 2 - 3 | 264 | 113 | |
NR_103465.1 | 2 - 3 | 264 | 113 | |||
GenBank mRNA | AK031489.1 | - | 203 | 113 | ||
AK031579.1 | - | 2 - 3 | 209 | 113 | ||
AK088996.1 | - | 2 - 3 | 194 | 113 | ||
AK145937.1 | - | 2 - 3 | 218 | 113 | ||
AK150000.1 | - | 2 - 3 | 240 | 113 | ||
AK153397.1 | - | 2 - 3 | 226 | 113 | ||
BC002056.1 | - | 2 - 3 | 192 | 3493119 | 113 | |
U08215.1 | - | 2 - 3 | 224 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |