Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163379.1 | NP_001156851.1 | 3 - 4 | 644 | 74 | |
NM_018862.3 | NP_061350.3 | 3 - 4 | 396 | 74 | ||
GenBank mRNA | AB005623.1 | - | 3 - 4 | 642 | 74 | |
AK086281.1 | - | 3 - 4 | 586 | 74 | ||
BC009651.1 | - | 3 - 4 | 396 | 3487719 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |