Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019800.4 | NP_062774.2 | 1 - 2 | 588 | 109 | |
GenBank mRNA | AB030038.1 | - | 1 - 2 | 581 | 109 | |
AF216223.1 | - | 1 - 2 | 65 | 109 | ||
AK002783.1 | - | 1 - 2 | 503 | 108 | ||
AK077729.1 | - | 1 - 2 | 576 | 109 | ||
AK160271.1 | - | 1 - 2 | 507 | 109 | ||
AK165356.1 | - | 1 - 2 | 589 | 109 | ||
AK171757.1 | - | 1 - 2 | 503 | 109 | ||
BC047276.1 | - | 1 - 2 | 328 | 6487551 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |