Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019933.2 | NP_064317.2 | 15 - 16 | 1537 | 76 | |
GenBank mRNA | AF106702.1 | - | 14 - 15 | 1365 | 76 | |
AK046683.1 | - | 15 - 16 | 1812 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mus musculus Probe spans exons |