Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022305.4 | NP_071641.1 | 3 - 4 | 1574 | 76 | |
GenBank mRNA | AK029474.1 | - | 3 - 4 | 827 | 76 | |
AK131721.1 | - | 3 - 4 | 677 | 76 | ||
AK132970.1 | - | 3 - 4 | 1575 | 76 | ||
AK154393.1 | - | 3 - 4 | 1033 | 76 | ||
AK167322.1 | - | 3 - 4 | 833 | 76 | ||
AK210987.1 | - | 1 - 2 | 204 | 76 | ||
AK214993.1 | - | 1 - 2 | 204 | 76 | ||
BC053006.1 | - | 3 - 4 | 1035 | 5698356 | 76 | |
D00314.1 | - | 3 - 4 | 988 | 76 | ||
J03880.1 | - | 3 - 4 | 1013 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |