Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164290.1 | NP_001157762.1 | 4 - 5 | 426 | 64 | |
NM_001164291.1 | NP_001157763.1 | 3 - 4 | 318 | 64 | ||
NM_001164292.1 | NP_001157764.1 | 3 - 4 | 315 | 64 | ||
NM_001164293.1 | NP_001157765.1 | 3 - 4 | 318 | 64 | ||
NM_001164294.1 | NP_001157766.1 | 3 - 4 | 318 | 64 | ||
NM_001164295.1 | NP_001157767.1 | 3 - 4 | 315 | 64 | ||
NM_024192.2 | NP_077154.2 | 4 - 5 | 417 | 64 | ||
GenBank mRNA | AB232359.1 | - | 2 - 3 | 216 | 64 | |
AK008842.1 | - | 3 - 4 | 281 | 64 | ||
AK013884.1 | - | 3 - 4 | 276 | 64 | ||
AK186231.1 | - | 3 - 4 | 241 | 64 | ||
AK207375.1 | - | 4 - 5 | 343 | 64 | ||
BC003442.1 | - | 4 - 5 | 381 | 2650719 | 64 | |
BY279028.1 | - | 3 - 4 | 317 | 64 | ||
BY312150.1 | - | 3 - 4 | 279 | 64 | ||
CK625575.1 | - | 3 - 4 | 244 | 64 | ||
CX223880.1 | - | 4 - 5 | 382 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |