Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025392.2 | NP_079668.2 | 5 - 6 | 629 | 55 | |
GenBank mRNA | AK003666.1 | - | 5 - 6 | 626 | 55 | |
AK010684.1 | - | 5 - 6 | 623 | 55 | ||
BC048465.1 | - | 5 - 6 | 621 | 6534043 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |