Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026011.3 | NP_080287.1 | 5 - 6 | 636 | 75 | |
GenBank mRNA | AB185206.1 | - | 5 - 6 | 440 | 75 | |
AK005999.1 | - | 5 - 6 | 483 | 75 | ||
AK009279.1 | - | 5 - 6 | 573 | 75 | ||
AK012024.1 | - | 5 - 6 | 600 | 75 | ||
AK030689.1 | - | 5 - 6 | 616 | 75 | ||
AK144177.1 | - | 5 - 6 | 607 | 75 | ||
AK146307.1 | - | 5 - 6 | 602 | 75 | ||
AK149822.1 | - | 5 - 6 | 586 | 75 | ||
AK151036.1 | - | 5 - 6 | 597 | 75 | ||
AK159311.1 | - | 4 - 5 | 283 | 75 | ||
AK159348.1 | - | 5 - 6 | 602 | 75 | ||
AK169929.1 | - | 5 - 6 | 570 | 75 | ||
AK170645.1 | - | 5 - 6 | 602 | 75 | ||
AK201340.1 | - | 3 - 4 | 191 | 75 | ||
AK206384.1 | - | 3 - 4 | 191 | 75 | ||
AK216939.1 | - | 1 - 2 | 71 | 75 | ||
AK219184.1 | - | 3 - 4 | 191 | 75 | ||
AK219830.1 | - | 3 - 4 | 191 | 75 | ||
BC013719.1 | - | 5 - 6 | 585 | 3983809 | 75 | |
CT010337.1 | - | 5 - 6 | 440 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |