Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165902.1 | NP_001159374.1 | 7 - 8 | 1168 | 75 | |
NM_007614.3 | NP_031640.1 | 7 - 8 | 1368 | 75 | ||
GenBank mRNA | AK020013.1 | - | 7 - 8 | 1284 | 75 | |
AK035311.1 | - | 7 - 8 | 1281 | 75 | ||
AK132398.1 | - | 7 - 8 | 1349 | 75 | ||
AK133656.1 | - | 7 - 8 | 1281 | 75 | ||
AK135743.1 | - | 4 - 5 | 835 | 75 | ||
AY751549.1 | - | 7 - 8 | 1293 | 75 | ||
BC043078.1 | - | 4 - 5 | 770 | 75 | ||
BC043481.1 | - | 3 - 4 | 402 | 75 | ||
BC048153.1 | - | 7 - 8 | 1293 | 5709247 | 75 | |
BC053065.1 | - | 7 - 8 | 1293 | 5709247 | 75 | |
M90364.1 | - | 7 - 8 | 1178 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Mouse Wnt Panel Mouse Stem Cell Pluripotency 1700 Inventoried Mouse_Stem_Cell Probe spans exons |