Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007664.4 | NP_031690.3 | 2 - 3 | 823 | 91 | |
GenBank mRNA | AB008811.1 | - | 2 - 3 | 500 | 91 | |
AK032826.1 | - | 2 - 3 | 263 | 91 | ||
AK146981.1 | - | 2 - 3 | 1020 | 91 | ||
BC022107.1 | - | 2 - 3 | 254 | 5100313 | 91 | |
M31131.1 | - | 2 - 3 | 500 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |